Learn More
Proteintech Group Inc Rat IL10 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE2000396T
Encompass_Preferred
Interleukin 10 is an essential anti-inflammatory cytokine that plays important roles as a negative regulator of immune responses to microbial antigens. IL10 is mainly secreted by macrophages, but also by T helper 1 (Th1) and Th2 lymphocytes, dendritic cells, cytotoxic T cells, B lymphocytes, monocytes and mast cells. It downregulates the expression of Th1 cytokines, MHC class II antigens, and co-stimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. IL10 can block NF-κB activity, and is involved in the regulation of the JAK-STAT signaling pathway. IL10 activity is mediated by the IL10 receptor (IL10R) which is a member of the class II cytokine receptor family.
Catalog No. 50-197-4175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.