Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Il17a Recombinant Protein

Catalog No. 89948681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-681 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-681 Supplier Abnova™ Supplier No. P4843
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: MAVLIPQSSVCPNAEANNFLQNVKVNLKVLNSLSSKASSRRPSDYLNRSTSPWTLSRNEDPDRYPSVIWEAQCRHQRCVNAEGKLDHHMNSVLIQQEILVLKREPEKCPFTFRVEKMLVGVGCTCVSSIVRHAS

Specifications

Accession Number Q61453
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 301289
Molecular Weight (g/mol) 30kDa
Name Il17a (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue
Quantity 25 μg
Immunogen MAVLIPQSSVCPNAEANNFLQNVKVNLKVLNSLSSKASSRRPSDYLNRSTSPWTLSRNEDPDRYPSVIWEAQCRHQRCVNAEGKLDHHMNSVLIQQEILVLKREPEKCPFTFRVEKMLVGVGCTCVSSIVRHAS
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Common Name Il17a
Gene Symbol Il17a
Biological Activity The activity is determined by the dose-dependent induction of IL-6 production in cultured mouse NIH 3T3 fibroblasts. The expected ED50 for this effect is 0.44-0.66ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.