Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Il1b Recombinant Protein

Catalog No. 89948747 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-747 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-747 Supplier Abnova™ Supplier No. P4841
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: VPIRQLHCRLRDEQQKCLVLSDPCELKALHLNGQNISQQVVFSMSFVQGETSNDKIPVALGLKGLNLYLSCVMKDGTPTLQLESVDPKQYPKKKMEKRFVFNKIEVKTKVEFESAQFPNWYISTSQAEHRPVFLGNSNGRDIVDFTMEPVSS

Specifications

Accession Number Q63264
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 24494
Molecular Weight (g/mol) 17.3kDa
Name Il1b (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue
Quantity 10 μg
Immunogen MVPIRQLHCRLRDEQQKCLVLSDPCELKALHLNGQNISQQVVFSMSFVQGETSNDKIPVALGLKGLNLYLSCVMKDGTPTLQLESVDPKQYPKKKMEKRFVFNKIEVKTKVEFESAQFPNWYISTSQAEHRPVFLGNSNGRDIVDFTMEPVSS
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Common Name Il1b
Gene Symbol Il1b
Biological Activity Recombinant rat Il1b is fully biologically active when compared to a standard. The ED50, as determined by the dose-dependent proliferation of mouse D10S cells, is 40.2pg/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.