Learn More
Proteintech Group Inc Rat IL2 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE2000296T
Encompass_Preferred
Interleukin-2 (IL2) is a protein initially identified as a T cell growth factor. IL2 is mainly produced by activated CD4+CD25- (helper) T cells, and induces the proliferation of these and others cells like CD8+ T cells, B and NK cells. For this reason, it has been used to treat immune-deficiencies like HIV, and induce immune response against tumors. Nevertheless, it has been shown that IL2 also acts as the main growth factor of CD4+CD25+ Regulatory T cells. Interleukin 2 mediates its functions in the target cell through the interleukin 2 receptor (IL2R). IL2R is a multimeric functional protein consisting of three different chains: Il2Rα, Il2Rβ and γc.
Catalog No. 50-197-4174
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.