Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat IL25 Recombinant Protein

Catalog No. 89948843 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-843 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-843 Supplier Abnova™ Supplier No. P4845
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. This cytokine can induce NF-kappaB activation, and stimulate the production of IL8. Both this cytokine and IL17B are ligands for the cytokine receptor IL17BR. Studies of the similar gene in mice suggested that this cytokine may be a proinflammatory cytokine favoring Th2-type immune response. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: CSHLPRCCPSKQQEFPEEWLKWNPAPVSPPEPLRHTHHPESCRASKDGPLNSRAISPWSYELDRDLNRVPQDLYHARCLCPHCVSLQTGSHMDPMGNSVPLYHNQTVFYRRPCHGEQGAHGRYCLERRLYRVSLACVCVRPRMMA

Specifications

Accession Number D3ZLB1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 64806
Molecular Weight (g/mol) 35.5kDa
Name IL25 (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel stained with Coomassie Blue
Quantity 10 μg
Immunogen MCSHLPRCCPSKQQEFPEEWLKWNPAPVSPPEPLRHTHHPESCRASKDGPLNSRAISPWSYELDRDLNRVPQDLYHARCLCPHCVSLQTGSHMDPMGNSVPLYHNQTVFYRRPCHGEQGAHGRYCLERRLYRVSLACVCVRPRMMA
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized 20 mM HCl, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias IL-17E/IL-25/IL17E
Common Name IL25
Gene Symbol IL25
Biological Activity The activity is determined by the dose-dependent induction of human IL-8 production from human PBMCs. The expected ED50 for this effect is 25-37.5ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.