Learn More
Sino Biological Rat IL2RG PAb 100UL 80197-RP01-100
Supplier:  Sino Biological 80197RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL2RG / CD132 (rR IL2RG / CD132; Catalog#80197-R08H; Q68FU6; Met1-Ala262). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.