Learn More
Sino Biological Rat IL4R MAb 25TESTS 80198-R002-A-25
Supplier:  Sino Biological 80198R002A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL4R (rR IL4R; Catalog#80198-R08H; Q63257-1; Met1-Arg232) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0584
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.