Learn More
Sino Biological Rat IL4R PAb 100UL 80198-T16-100
Supplier:  Sino Biological 80198T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-4R / CD124 (rR IL-4R / CD124; Catalog#80198-R08H; Q63257-1; Met1-Arg232). IL-4R / CD124 specific IgG was purified by Rat IL-4R / CD124 affinity chromatography.
Catalog No. 50-241-0563
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.