Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Il6 Recombinant Protein

Catalog No. 89948939 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-948-939 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-948-939 Supplier Abnova™ Supplier No. P4842
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: MFPTSQVRRGDFTEDTTHNRPVYTTSQVGGLITYVLREIFEMRKELCNGNSDCMNSDDALSENNLKLPEIQRNDGCFQTGYNQEICLLKICSGLLEFRFYLEFVKNNLQDNKKDKARVIQSNTETLVHIFKQEIKDSYKIVLPTPTSNALLMEKLESQKEWLRTKTIQLILKALEEFLKVTMRSTRQT

Specifications

Accession Number P20607
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 24498
Molecular Weight (g/mol) 21.7kDa
Name Il6 (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MFPTSQVRRGDFTEDTTHNRPVYTTSQVGGLITYVLREILEMRKELCNGNSDCMNSDDALSENNLKLPEIQRNDGCFQTGYNQEICLLKICSGLLEFRFYLEFVKNNLQDNKKDKARVIQSNTETLVHIFKQEIKDSYKIVLPTPTSNALLMEKLESQKEWLRTKTIQLILKALEEFLKVTMRSTRQT
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 10 mM HCl, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias ILg6/Ifnb2
Common Name Il6
Gene Symbol Il6
Biological Activity The activity is determined by the dose-dependent stimulation of mouse 7TD1 cells. The expected ED50 for this effect is 5.9-8.9pg/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.