Learn More
Biomatik Corporation Rat Interleukin 34 (IL34) CLIA Kit
Supplier:  Biomatik Corporation EKU09805
Encompass_Preferred
Rat Interleukin 34 (IL34) CLIA Kit for detecting IL34 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.59pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q4KM46. Gene ID: 498951. Target Synonyms: C16orf77. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5237
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.