Learn More
Biomatik Corporation Rat Kelch Repeat And BTB Domain Containing Protein 10 (KBTBD10) ELISA Kit
Supplier:  Biomatik Corporation EKU12381
Encompass_Preferred
Rat Kelch Repeat And BTB Domain Containing Protein 10 (KBTBD10) ELISA Kit for detecting KBTBD10 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ER30. Gene ID: 117537. Target Synonyms: KRP1; KBTBD10; Sarcosin; Sarcomeric Muscle Protein; Kelch-like protein 41; Kel-like protein 23; Kelch-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.