Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat LILRA5 (CD85f, LIR-9) Fc Chimera Recombinant Protein

Catalog No. PIRP88272
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88272 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88272 Supplier Invitrogen™ Supplier No. RP88272
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

Leukocyte immunoglobulin-like receptor A5 (LILRA5) is a gene located on chromosome 19q13.42, encoding a protein that is part of the leukocyte immunoglobulin-like receptor (LILR) family. These receptors are primarily expressed on immune cells, such as natural killer cells, monocytes, and dendritic cells, and play a pivotal role in modulating immune responses. LILRA5 is thought to function as an activating receptor, influencing the activation and regulation of immune cell functions. It is implicated in processes such as antigen presentation, cytokine production, and phagocytosis, contributing to both innate and adaptive immune responses. Dysregulation of LILRA5 expression or function can impact immune homeostasis and has been associated with various autoimmune and inflammatory diseases. Research into LILRA5 aims to uncover its specific ligands and signaling pathways, with potential implications for therapeutic targeting in immune-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 691533
Molecular Weight (g/mol) 63 kDa
Name Rat LILRA5 (CD85f, LIR-9) Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD85; CD85 antigen-like family member F; CD85f; CD85f antigen; E030017K20; EG232801; Gm4878; ILT11; ILT-11; Immunoglobulin-like transcript 11; immunoglobulin-like transcript 11 protein; leucocyte Ig-like receptor A5; leukocyte Ig-like receptor 9; leukocyte immunoglobulin like receptor A5; leukocyte immunoglobulin-like receptor; leukocyte immunoglobulin-like receptor 9; Leukocyte immunoglobulin-like receptor subfamily A member 5; leukocyte immunoglobulin-like receptor subfamily A member 5 soluble; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 7; LILRA5; LILRB7; Lilrc1; LIR9; LIR-9
Common Name LILRA5
Gene Symbol LILRA5
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Rat LILRA5, amino acids Met1-Asn248 (Accession # D4A6Y0) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.