Learn More
American Research Products Inc Rat Mast Cell Tryptase MCT EL
Supplier:  American Research Products Inc CSBE13627R
Encompass
Rat Mast Cell Tryptase MCT ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma urine Method Sandwich
Catalog No. 50-003-28274