Learn More
Biotang Inc Rat matrix metalloproteinase 9/Gelatinase B,MMP-9 ELISA Kit , 96T
Supplier:  Biotang Inc R6566
Encompass_Preferred
Rat matrix metalloproteinase 9/Gelatinase B,MMP-9 ELISA Kit , 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-155-276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.