Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Rat/Mouse Cytochrome c Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat Cytochrome c in cell lysates (50 ul), subcellular fractions (50 ul).
Specifications
Specifications
| Assay Range | 0.78 - 25ng/mL |
| Assay Sensitivity | 0.5ng/mL |
| Conjugate | HRP |
| Sample Type | Cell Lysates, Subcellular Fractions |
| Specificity | Reduced and oxidized forms of natural Rat and Mouse cytochrome c |
| For Use With (Application) | ELISA |
| Quantity | 1 x 96-well Plate |
| Sample Volume | Cell Lysates (50uL),Subcellular Fractions (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | CYCHCS, CYCS, cytochrome c, cytochrome c, somatic, THC4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
