Learn More
Mercodia Inc RAT/MOUSE PROINSULIN ELISA KIT
SDP
Supplier:  Mercodia Inc 10123201
Encompass_Preferred
Mercodia Rat/Mouse Proinsulin ELISA provides a method for the quantitative determination of proinsulin in rat and mouse serum, plasma, cell culture medium and cellular extracts. Assay is specific for rat and mouse proinsulin I and II. No cross-reactivity to insulin or c-peptide. Fast and easy-to-use.
Catalog No. NC9088881
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.