Learn More
Specifications
Specifications
| Description | Paraffin Tissue Section Panel - Rat Normal Tissue, Multitissue Set I mounted on positively charged glass slides |
| Quantity | 5 Slides |
| Species | Rat |
| Tissue Condition | Normal |
| Type | Multiple |
| For Use With (Application) | Paraffin Tissue Section Panel - Rat Normal Tissue, Multi-tissue I |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
