Learn More
Biomatik Corporation Rat Myosin Heavy Chain 8, Skeletal Muscle, Perinatal (MYH8) ELISA Kit
Supplier:  Biomatik Corporation EKU06103
Encompass_Preferred
Rat Myosin Heavy Chain 8, Skeletal Muscle, Perinatal (MYH8) ELISA Kit for detecting MYH8 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.50-4000pg/ml. Sensitivity: 24.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04462. Target Synonyms: MyHC-peri; MyHC-pn; MyHC-perinatal; Myosin heavy chain, skeletal muscle, perinatal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.