Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat NKp30/NCR3 (NP_861543) VersaClone cDNA

Catalog No. RDC1905
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1905 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1905 Supplier R&D Systems™ Supplier No. RDC1905
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Rat NKp30/NCR3 (NP_861543) VersaClone cDNA is a member of the natural killer receptor family.

The Rat NKp30/NCR3 (NP_861543) VersaClone cDNA clone contains the complete ORF for NKp30/NCR3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The NKp30/NCR3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Specifications

Gene NKp30/NCR3
Accession Number NP_861543
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 601bp
pH 8.5
Gene ID (Entrez) 259197
Gene Alias Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Rat
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.