Learn More
Sino Biological Rat NTRK1 PAb 100UL 80404-T24-100
Supplier:  Sino Biological 80404T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat TrkA / NTRK1 (Catalog#80404-R08H; NP_067600.1; Met1-Pro418). TrkA / NTRK1 specific IgG was purified by Rat TrkA / NTRK1 affinity chromatography.
Catalog No. 50-240-8291
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.