Learn More
Novus Biologicals™ Rat p21/CIP1/CDKN1A ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275207
Description
The Rat p21/CIP1/CDKN1A ELISA Kit (Colorimetric) measures rat p21/CIP1/CDKN1A in serum, plasma and other biological fluids.Specifications
78.13 - 5000 pg/mL | |
This kit recognizes Rat CDKN1A in samples. No significant cross-reactivity or interference between Rat CDKN1A and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1026 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
46.88 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CDKN1A | |
1 Kit | |
p21/CIP1/CDKN1A |