Learn More
Biomatik Corporation Rat Phospholipase A2, Group IIA (PLA2G2A) ELISA Kit
Supplier:  Biomatik Corporation EKU06615
Encompass_Preferred
Rat Phospholipase A2, Group IIA (PLA2G2A) ELISA Kit for detecting PLA2G2A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.188-12ng/ml. Sensitivity: 0.082ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14423. Gene ID: 29692. Target Synonyms: PLA2; MOM1; PLA2B; PLA2L; PLA2S; PLAS1; sPLA2 (platelets,synovial fluid) ; Non-pancreatic secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.