Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Rat PlGF-1 Recombinant Protein, PeproTech®

Catalog No. 400395UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
400-39-5UG 5 μg
40039100UG 100 μg
400-39-1MG 1 mg
40039250UG 250 μg
400-39-25UG 25 μg
40039500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 400-39-5UG Supplier Gibco™ Supplier No. 400395UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Rat PlGF-1 is a 30.3 kDa disulfide-linked homodimeric protein of two 135 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q63434
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 94203
Molecular Weight (g/mol) 30.3 kDa
Name Rat PlGF-1
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Placenta Growth Factor; Placenta Growth Factor-1
Common Name PlGF-1
Gene Symbol PGF
Conjugate Unconjugated
Recombinant Recombinant
Sequence ALSAGNNSTE MEVVPFNEVW GRSYCRPMEK LVYIADEHPN EVSHIFSPSC VLLSRCSGCC GDEGLHCVAL KTANITMQIL KIPPNRDPHS YVEMTFSQDV LCECRPILET TKAERRKTKG KRKQSKTPQT EEPHL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.