Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Prl (P01237, 198 amino acids) Partial Recombinant Protein

Catalog No. 89958246 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-246 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-246 Supplier Abnova™ Supplier No. P5144

Used for Func, SDS-PAGE

Specifications

Accession Number P01237
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 24683
Molecular Weight (g/mol) 22.5kDa
Name Prl (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 2 ug/lane in SDS-PAGE
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias PRLB/PRLSD1/Prl1a1/Prol/RATPRLSD1/RNPROL
Common Name Prl
Gene Symbol Prl
Biological Activity The ED50 determined by the dose-dependent stimulation of the proliferation of rat lymphoma, Nb2-11 cells was found to be in the range of 0.1-1.0ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.