Learn More
Kingfisher Biotech Inc Rat Prolactin Receptor Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2030R500
Encompass_Preferred
The Rat Prolactin Receptor yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rat Prolactin Receptor applications are for cell culture, ELISA standard, and Western Blot Control. Rat Prolactin Receptor yeast-derived recombinant protein can be purchased in multiple sizes. Rat Prolactin Receptor Specifications: (Molecular Weight: 24.6 kDa) (Amino Acid Sequence: QSPPGKPEIH KCRSPDKETF TCWWNPGTDG GLPTNYSLTY SKEGEKTTYE CPDYKTSGPN SCFFSKQYTS IWKIYIITVN ATNQMGSSSS DPLYVDVTYI VEPEPPRNLT LEVKQLKDKK TYLWVKWSPP TITDVKTGWF TMEYEIRLKP EEAEEWEIHF TGHQTQFKVF DLYPGQKYLV QTRCKPDHGY WSRWSQESSV EMPNDFTLKD (210)) (Gene ID: 24684). For research use only. Made in the USA
Catalog No. 50-253-2511
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.