Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Prolactin ELISA Kit

Catalog No. 502545420
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-5420 10 x 96 Tests
ERA50RBX5 5 x 96 Tests
ERA50RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-254-5420 Supplier Invitrogen™ Supplier No. ERA50RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Rat Prolactin quantitates rat Prolactin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Prolactin.

Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL
Assay Range 0.41 to 100 ng/mL
Assay Sensitivity 0.41 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 24683
Gene Symbol PRL
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Prolactin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.