Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Pure-Blot anti-Mouse IgG (H+L) Secondary Antibody (eB144), Biotin, Novus Biologicals™
SDP

Catalog No. NB120515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120515 Supplier Novus Biologicals Supplier No. NBP311660
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG Monoclonal antibody specifically detects IgG in Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IgG
Applications ELISA, Western Blot
Classification Monoclonal
Clone eB144
Conjugate Biotin
Dilution Western Blot 1:000, Immunoprecipitation
Formulation 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 (Proprietary Stabilizer)
Gene Alias IP Detection Reagent
Host Species Rat
Immunogen Mouse IgG
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.