Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Reelin ELISA Kit (Colorimetric)
SDP

Catalog No. NB030432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB030432 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB030432 Supplier Novus Biologicals™ Supplier No. NBP282503
Only null left
Add to Cart
Add to Cart

Rat Reelin ELISA Kit (Colorimetric) measures Rat Reelin in Serum, plasma, cell supernatant, and other biological fluids.

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Rat Reelin in samples. No Significant cross-reactivity or interference between Rat Reelin and analogues was observed.
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 5649
Gene Symbol RELN
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 3.4.21, EC 3.4.21.-, reelin, RLPRO1598
Target Reelin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.