Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Retn (NP_653342, 94 amino acids) Partial Recombinant Protein with His tag

Catalog No. 89959769 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-769 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-959-769 Supplier Abnova™ Supplier No. P5141

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used for SDS-PAGE

Specifications

Accession Number NP_653342
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 246250
Molecular Weight (g/mol) 11.9kDa
Name Retn (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 25 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Common Name Retn
Gene Symbol Retn
Species E. coli
Recombinant Recombinant
Protein Tag His
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.