Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat RTN1-A/NSP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5996SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5996SP 25 μg
MAB5996 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5996SP Supplier R&D Systems Supplier No. MAB5996SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

RTN1-A/NSP Monoclonal specifically detects RTN1-A/NSP in Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen RTN1-A/NSP
Applications Immunohistochemistry
Classification Monoclonal
Clone 559706
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q64548
Gene Alias NSP, NSP-A, RTN1, RTN1A
Gene Symbols Rtn1
Host Species Mouse
Immunogen E. coli-derived recombinant rat RTN1-A/NSP Met73-Ser502 Accession # Q64548
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6252
Test Specificity Detects rat RTN1-A/NSP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant rat RTN-A2B, recombinant human (rh) RTN1-A, and rhRTN3-A1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.