Learn More
Specifications
Specifications
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 37.5 pg/mL |
| Detection Method | Chemiluminescence |
| Sample Type | Serum, Plasma, Other Biological Fluids |
| Specificity | This kit recognizes Rat RUNX2/CBFA1 in samples. No significant cross-reactivity or interference between Rat RUNX2 and analogues was observed |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 860 |
| Gene Symbol | RUNX2 |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
