Learn More
Biomatik Corporation Rat Sequestosome 1 (SQSTM1) ELISA Kit
Supplier:  Biomatik Corporation EKU11737
Encompass_Preferred
Rat Sequestosome 1 (SQSTM1) ELISA Kit for detecting SQSTM1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08623. Gene ID: 113894. Target Synonyms: OSIL; PDB3; ZIP3; p62; p62B; p60; A170; Paget Disease Of Bone 3; Oxidative Stress Induced Like; EBI3-associated protein of 60 kDa; Ubiquitin-binding protein p62. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7154
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.