Learn More
GIBCO™ Rat (Sprague Dawley) Cytosol
Description
Liver cytosolic fractions prepared from a pool of male Sprague Dawley rats.
Total protein concentration: 20 mg/ml
We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.
For research use only. Not intended for human or animal therapeutic or diagnostic use.
Related Link
Learn more about our microsomes and subcellular fractions
Specifications
Specifications
| Quantity | 1 mL |
| Purity | Research Grade |
| Host Species | Rat |
| Concentration | 20 mg/mL |
| Product Type | Rat Cytosol |
| Content And Storage | Store between -60°C and -85°C. |
| Format | Tube |
| Sub Cellular Localization | Cytosol |
| Shipping Condition | Dry Ice |
| Product Line | Gibco |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.