Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIBCO™ Rat (Sprague Dawley) S9 Fractions

Catalog No. RTS9PL
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RTS9PL 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RTS9PL Supplier GIBCO™ Supplier No. RTS9PL
  • $75.75
    $55.65 / Each of 1

    Save $20.10

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Liver S9 fractions prepared from a pool of male Sprague Dawley rats. Total protein concentration: 20mg/mL

Liver S9 fractions prepared from a pool of male Sprague Dawley rats.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions

TRUSTED_SUSTAINABILITY

Specifications

Gender Male
Physical Form Frozen
Tissue Liver
Donor Source Pooled
Quantity 1 mL
Purity Research Grade
Host Species Rat
Concentration 20 mg/mL
Product Type Rat S9 Fraction
Content And Storage Store between -60°C and -85°C.
Format Tube
Sub Cellular Localization S9 Fraction
Shipping Condition Dry Ice
Product Line Gibco
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.