Learn More
Innovative Research Inc Rat Sprague Dawley Red Blood Cells 25ml
Supplier:  Innovative Research Inc IRTSDRBC25ML
Encompass_Preferred
Rat Sprague Dawley Red Blood Cells from Innovative Research are manufactured using whole blood collections from type Sprague Dawley rats. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. Red blood cells are perishable and are collected and processed upon receipt of order. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4871
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.