Learn More
American Research Products Inc Rat Thyroid PeroXidase TPO EL
Supplier:  American Research Products Inc CSBE08352R
Encompass
Rat Thyroid-PeroXidase TPO ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-40ng/mL Sensitivity 2 5 ng/mL Sample serum plasma tissue Homogenates Method Competitive
Catalog No. 50-003-28940