Learn More
Description
- Also known as: Kim1; KIM-1
- TIM-1 is a type I transmembrane glycoprotein member of the TIM family of immunoglobulin superfamily molecules.
- This family is involved in regulation of Th1 and Th2-cell-mediated immunity.
- TIM-1 is a receptor for TIMD4 and the hepatitis A virus.
- TIM-1 ligation induces T cell proliferation and promotes cytokine production. TIM1 may be involved in the moderation of asthma and allergic diseases.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | rHavcr1 |
| Accession Number | NP_775172 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 937 bp |
| Gene ID (Entrez) | 26762 |
| Gene Alias | CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Rat |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
