Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat TIMP-2 ELISA Kit

Catalog No. ERA54RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ERA54RB 96 Tests
ERA54RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ERA54RB Supplier Invitrogen™ Supplier No. ERA54RB
Add to Cart
Add to Cart

ELISA

Rat TIMP-2 quantitates rat TIMP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat TIMP-2.

TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72 kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number collagenase inhibitor; CSC-21 K; D11Bwg1104e; DDC8; Metalloproteinase inhibitor 2; metalloproteinase inhibitor precursor; metalloproteinase inhibitor TIMP-2; similar to metalloproteinase inhibitor; testicular secretory protein Li 57; TIMP 2; TIMP metallopeptidase inhibitor 2; Timp2; TIMP-2; TIMP-2 protein; tissue inhibitor of matrix metalloproteinase-2; tissue inhibitor of metalloproteinase 2; tissue inhibitor of metalloproteinase-2; tissue inhibitor of metalloproteinases 2; tissue inhibitor of mettaloproteinase 2
Assay Range 20 to 5,000 pg/mL
Assay Sensitivity 20 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 29543
Gene Symbol TIMP2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TIMP2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.