Learn More
Novus Biologicals™ Rat TRACP/PAP/ACP5 ELISA Kit (Chemiluminescence)
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 125 to 8000 pg/mL |
| Assay Sensitivity | 75 pg/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat TRACP-5b in samples. No significant cross-reactivity or interference between Rat TRACP-5b and analogues was observed. |
| Gene ID (Entrez) | 54 |
| Gene Symbol | ACP5 |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.