Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Vegfc (P97953, 129 amino acids) Partial Recombinant Protein

Catalog No. 89967017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-017 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-967-017 Supplier Abnova™ Supplier No. P5143

Used for Func, SDS-PAGE

Specifications

Accession Number O35757
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 114111
Molecular Weight (g/mol) 18.0kDa
Name Vegfc (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with 0.1% acetic acid to a concentration of 0.1-1.0 mg/mL, store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg
Gene Alias AW228853, VEGF-C
Common Name Vegfc
Gene Symbol Vegfc
Biological Activity Activity of Vegfc will be tested by the stimulation of VEGF-R3 phosphorylation in cells transfected with VEGF-R3. The expected ED50 is > 200ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.