Learn More
R&D Systems™ Rat VisUCyte™ HRP Polymer-DAB Cell and Tissue Staining Kit
Shop All Novus Biologicals Products

Description
R&D Systems™ VisUCyte™ Cell and Tissue Staining Kits are intended for localization of antigens in a broad range of histological and cytological specimens. Detection is based on a secondary antibody and HRP enzyme conjugated to a polymer backbone for detection of primary antibodies that react with tissue antigens under study. Visualization is based on enzymatic conversion of a chromogenic substrate 3,3′ Diaminobenzidine (DAB) into a colored brown precipitate by horseradish peroxidase (HRP) at the sites of antigen localization, which can then be viewed using bright- field microscopy.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2-8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 50 Tests |
| Species | Rat |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.