Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ravulizumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542134
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542134 Supplier Invitrogen™ Supplier No. MA542134
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Ravulizumab is considered a long-acting complement 5 (C5) inhibitor that has been undergoing clinical trials for the treatment of paroxysmal nocturnal hemoglobinuria (PNH) as of 4 February, 2016. A pharmaceutical similar to ravulizumab (ALXN1210), called eculizumab, is currently approved for the treatment of PNH in 46 countries under the brand name Soliris™. Ravulizumab is considered by Alexion Pharmaceuticals Inc. to be a 'next-generation' eculizumab molecule. Ravulizumab was subsequently approved by the US FDA in December of 2018 for a variety of beneficial characteristics that make it an advanced, next-generation agent in comparison to eculizumab. In particular, ravulizumab is currently the first and only long-acting C5 complement inhibitor that can be administered every eight weeks for the treatment of adult patients with PNH whereas eculizumab is a bi-weekly treatment, Label. Moreover, virtually all of the phase 3 trial results for ravulizumab have demonstrated the equivalent efficacy and safety established by eculizumab and that patients transition safely and effectively from using eculizumab to ravulizumab. Subsequently, whereas PNH patients may have needed to previously plan their lives rather strictly around the bi-weekly infusion administrations of eculizumab, with ravulizumab such patients can find a more relaxed dosing schedule of only six or seven infusions over an entire year, Label.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ravulizumab Humanized
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias ALXN-1210; ravulizumab-cwvz
Host Species Human
Immunogen Human C5.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2 κ, IgG4 κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.