Learn More
Specifications
Specifications
| For Use With (Application) | Western Blotting |
| Product Type | Raw 264.7 Whole Cell - T0901317 treated Assay Kit - (Dry Ice) |
| Content And Storage | NBP1-30157: Raw 264.7 Whole Cell Lysate, NBP1-30158: Raw 264.7 Whole Cell Lysate. Store at -20°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Quantity | 2 Vials |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
