Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAX Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-517 Supplier Invitrogen™ Supplier No. PA141388
  • $587.00
    $487.70 / Each of 1

    Save $99.30

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: Jurkat whole cell lysate. Reacts with New World Monkey. 85% sequence identity with chicken and 78% with zebrafish, mouse and rat proteins.

Rax plays a critical role in eye formation by regulating the initial specification of retinal cells and/or their subsequent proliferation. It binds to the photoreceptor conserved element-I (PCE-1/Ret 1) in the photoreceptor cell-specific arrestin promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene RAX
Gene Accession No. Q9Y2V3
Gene Alias 7530406A22Rik; E130303K03Rik; ey1; ey-1; eyeless 1; LOW QUALITY PROTEIN: retinal homeobox protein Rx; MCOP3; RAX; retina and anterior neural fold homeobox; retina and anterior neural fold homeobox protein; Retinal homeobox protein Rx; Rx; Rx/rax homeoprotein
Gene Symbols RAX
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human Rax.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30062, 610136
Target Species Human, Canine, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.