Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Ray Box
SCI_ED

Catalog No. S27889 Shop All Eisco Products
Change view
Click to view available options
Grade:
5 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27889 5 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27889 Supplier Eisco™ Supplier No. RBX14
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A simple, safe, and sturdy source of projected light for many classic optics experiments.

  • Perfect for use with acrylic or glass demonstration optics to teach geometrical optics
  • Ray box produces parallel convergent rays across a horizontal plane with various configurations to create different ray patterns
  • Does not require a completely dark room to conduct optic experiments due to high intensity light used
  • Works off of standard 100 to 240 V wall power, no power supply required (although one can be used for more power)
  • Supplied with a multi slit that produces either a single wide beam or choice of one, three, or five pencil thin rays
  • Apparatus Includes: (1) Sheet Metal Outer Case, (1) Halogen Bulb, (1) Fixed Lens, (1) Wall Plug, (1) Skit Card with Multiple Options
  • Built in solid aluminum extrusion casing

Specifications

Product Type Ray Box
Description Halogen Source
Grade 5 to 14
Material Aluminum
For Use With (Application) Optics Experiments
Includes Sheet Metal Outer Case, Halogen Bulb, Fixed Lens, Wall Plug, and Skit Card with Multiple Options
Electrical Requirements 100 to 240 V, 50/60 Hz
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.