Learn More
Tritech Forensics Inc RAY REAGENT 500ML
SDP
Supplier:  Tritech Forensics Inc PF30104500ML
Encompass
RAY dye stain is a fluorescent mixture of Rhodamine, Ardrox, and Basic Yellow. Use RAY with a forensic light source after processing with cyanoacrylate. Particularly useful in the enhancement of CA developed prints on plastic bags. RAY enhanced latent prints are visualized between 415nm and 530nm, making it ideal for use with a wide variety of forensic light sources.
Catalog No. NC2583426
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.