Learn More
West Chester™ Extra Heavy Weight Cotton Hot Mill Gloves with Two-Layers of Rayon Lining
Catalog No. 19821576 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Large
Catalog No.
19-821-576 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-821-576 Supplier West Chester™ Supplier No. G03SI
This item is not returnable. View return policy
Cotton construction offers comfort, insulation, and superior heat protection while rayon lining offers provides good heat dispersion. These gloves can be used in forging, hot metal handling, castings, molds, and other operations that require heat protection.
  • Clute cut provides a roomy fit
  • Nap-out finish for heat resistance
  • Knuckle strap for extra protection
  • Straight thumb increases comfort in closed-fisted work
  • Gauntlet cuff for added forearm protection, allows for fast removal
Specifications
Description Extra Heavy Weight
Exterior Finish Nap-out