Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB705 Hamster Anti-Mouse CD11c (Integrin αX), BD Horizon™

Catalog No. BDB570714 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570714 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570714 Supplier BD Biosciences Supplier No. 570714
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

CD11c (Integrin αX) Monoclonal Antibody Specifically detects CD11c (Integrin αX) in Mouse Samples. It is validated for Flow Cytometry.

The N418 monoclonal antibody specifically binds to CD11c, which is the Integrin alpha X (Integrin αX, Itgax) chain of the heterodimeric gp150, 95 (CD11c/CD18, αXβ2) integrin that forms the complement receptor 4 (CR4). CD11c is a 150 kDa type I transmembrane glycoprotein found on dendritic cells, CD4-CD8+ intestinal intraepithelial lymphocytes (IEL), and some NK cells and T cells. Its expression increases on IEL and T cells after activation. Monocyte/macrophage lineage cells have also been reported to express low levels of CD11c. The CD11c/CD18 integrin can bind to ligands such as iC3b, fibrinogen, and CD54, and is involved in phagocytosis and cellular interactions during inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c (Integrin αX)
Applications Flow Cytometry
Classification Monoclonal
Clone N418
Concentration 0.2 mg/mL
Conjugate RB705
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4
Host Species Armenian Hamster
Immunogen Mouse Dendritic Cells
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.