Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Rat IgG2a λ Isotype Control
Isotype Control
Supplier: BD Biosciences 570170
Description
It is validated for Flow cytometry|Isotype control
Specifications
Flow Cytometry, Control | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Trinitrophenyl–Keyhole Limpet Hemocyanin | |
0.1 mg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a λ |
B39-4 | |
RB780 | |
Anti-Keyhole Limpet Hemocyanin (KLH) | |
Affinity Chromatography | |
RUO | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction