Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBBP9 Monoclonal Antibody (OTI4G3)
GREENER_CHOICE

Catalog No. PIMA525239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525239 Supplier Invitrogen™ Supplier No. MA525239
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBBP9 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RBBP9
Gene Accession No. O75884
Gene Alias B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807
Gene Symbols RBBP9
Host Species Mouse
Immunogen Full length human recombinant protein of RBBP9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10741
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.